missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VPS29 Protein
A cDNA sequence encoding the VPS29 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13945-50ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
VPS29 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMMLVLV LGDLHIPHRC NSLPAKFKKL LVPGKIQHIL CTGNLCTKES YDYLKTLAGD VHIVRGDFDE NLNYPEQKVV TVGQFKIGLI HGHQVIPWGD MASLALLQRQ FDVDILISGH THKFEAFEHE NKFYINPGSA TGAYNALETN IIPSFVLMDI QASTVVTYVY QLIGDDVKVE RIEYKKP | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The VPS29 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 40% glycerol. |