missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VPS26A Protein
A cDNA sequence encoding the VPS26A was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13942-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
9559 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSFLGGF FGPICEIDIV LNDGETRKMA EMKTEDGKVE KHYLFYDGES VSGKVNLAFK QPGKRLEHQG IRIEFVGQIE LFNDKSNTHE FVNLVKELAL PGELTQSRSY DFEFMQVEKP YESYIGANVR LRYFLKVTIV RRLTDLVKEY DLIVHQLATY PDVNNSIKME VGIEDCLHIE FEYNKSKYHL KDVIVGKIYF LLVRIKIQHM ELQLIKKEIT GIGPSTTTET ETIAKYEIMD GAPVKGESIP IRLFLAGYDP TPTMRDVNKK FSVRYFLNLV LVDEEDRRYF KQQEIILWRK APEKLRKQRT NFHQRFESPE SQASAEQPEM | |
Greater than 85% as determined by SDS-PAGE. |
VPS26A Protein | |
Research Use Only | |
VPS26A | |
Human | |
His | |
VPS26A protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.15M NaCl. |