missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UPRT Protein
A cDNA sequence encoding the UPRT was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Name | UPRT Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15985069
|
enQuireBio™
QP13902-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15965069
|
enQuireBio™
QP13902-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15975069
|
enQuireBio™
QP13902-1MG |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
UPRT Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UPRT protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMATELQC PDSMPCHNQQ VNSASTPSPE QLRPGDLILD HAGGNRASRA KVILLTGYAH SSLPAELDSG ACGGSSLNSE GNSGSGDSSS YDAPAGNSFL EDCELSRQIG AQLKLLPMND QIRELQTIIR DKTASRGDFM FSADRLIRLV VEEGLNQLPY KECMVTTPTG YKYEGVKFEK GNCGVSIMRS GEAMEQGLRD CCRSIRIGKI LIQSDEETQR AKVYYAKFPP DIYRRKVLLM YPILSTGNTV IEAVKVLIEH GVQPSVIILL SLFSTPHGAK SIIQEFPEIT ILTTEVHPVA PTHFGQKYFG TD | |
Greater than 90.0% as determined by SDS-PAGE. |