missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TSLPR Protein

A cDNA sequence encoding the TSLPR was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13827-HIS-2ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15964829

Menge Preis
1 128.22 CHF / 2 Mikrogramm
10 117.77 CHF / 2 Mikrogramm
Voraussichtliches Versanddatum: 28-10-2021

Zum Warenkorb hinzufügen


Beschreibung

Beschreibung

Spezifikation

Spezifikation

64109
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
CRLF2
Human
His
TSLPR protein solution (1 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4M UREA and 10% glycerol.
TSLPR Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSQGGAAEG VQIQIIYFNL ETVQVTWNAS KYSRTNLTFH YRFNGDEAYD QCTNYLLQEG HTSGCLLDAE QRDDILYFSI RNGTHPVFTA SRWMVYYLKP SSPKHVRFSW HQDAVTVTCS DLSYGDLLYE VQYRSPFDTE WQSKQENTCN VTIEGLDAEK CYSFWVRVKA MEDVYGPDTY PSDWSEVTCW QRGEIRDACA ETPTPPKPKL SK
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate