missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Trefoil factor 2 Protein
A DNA sequence encoding the Human Trefoil factor 2, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10179-ec-250ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_005414.1 | |
Trefoil factor 2 Protein | |
Research Use Only | |
TFF2 | |
The ED50 as determined by a chemotaxis bioassay using human MCF-7 cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized |
7032 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
EKPSPCQCSR LSPHNRTNCG FPGITSDQCF DNGCCFDSSV TGVPWCFHPL PKQESDQCVM EVSDRRNCGY PGISPEECAS RKCCFSNFIF EVPWCFFPKS VEDCHY | |
Greater than 97% as determined by SDS-PAGE and HPLC. |