missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TPSAB1 Protein
A cDNA sequence encoding the TPSAB1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Name | TPSAB1 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15964739
|
enQuireBio™
QP13798-HIS-INSECT-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15944739
|
enQuireBio™
QP13798-HIS-INSECT-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15954739
|
enQuireBio™
QP13798-HIS-INSECT-1MG |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
TPSAB1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TPSAB1 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
IVGGQEAPRS KWPWQVSLRV HGPYWMHFCG GSLIHPQWVL TAAHCVGPDV KDLAALRVQL REQHLYYQDQ LLPVSRIIVH PQFYTAQIGA DIALLELEEP VNVSSHVHTV TLPPASETFP PGMPCWVTGW GDVDNDERLP PPFPLKQVKV PIMENHICDA KYHLGAYTGD DVRIVRDDML CAGNTRRDSC QGDSGGPLVC KVNGTWLQAG VVSWGEGCAQ PNRPGIYTRV TYYLDWIHHY VPKKPHHHHH H | |
Greater than 90.0% as determined by SDS-PAGE. |