missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFRSF12A / FN14 / TWEAKR Protein
A DNA sequence encoding the Human TNFRSF12A / FN14 / TWEAKR, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10331-ec-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_057723.1 | |
TNFRSF12A / FN14 / TWEAKR Protein | |
Research Use Only | |
TNFRSF12A | |
The ED50 as determined by inhibiting TWEAK- dependent proliferation of human umbilical vein endothelial cells (HUVEC) is less than 30 ng/ml, corresponding to a specific activity of > 3.3 M 104 IU/mg, in the presence of 15 ng/ml of rHuTWEAK. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
51330 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
EQAPGTAPCS RGSSWSADLD KCMDCASCRA RPHSDFCLGC AAAPPAPFRL LWP | |
Greater than 95% as determined by SDS-PAGE and HPLC. |