missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFR2 / CD120b / TNFRSF1B Protein
A DNA sequence encoding the Human TNFR2 / CD120b / TNFRSF1B, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10375-ec-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001057.1 | |
TNFR2 / CD120b / TNFRSF1B Protein | |
Research Use Only | |
TNFRSF1B | |
The ED50 as determined by its ability to inhibit the TNF-alpha mediated cytotoxicity in the L-929 cells is less than 0.2 ug/ml, corresponding to a specific activity of > 5000 IU/mg in the presence of 0.25 ng/mL of rHuTNF-alpha. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
7133 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MPAQVAFTPY APEPGSTCRL REYYDQTAQM CCSKCSPGQH AKVFCTKTSD TVCDSCEDST YTQLWNWVPE CLSCGSRCSS DQVETQACTR EQNRICTCRP GWYCALSKQE GCRLCAPLRK CRPGFGVARP GTETSDVVCK PCAPGTFSNT TSSTDICRPH QICNVVAIPG NASMDAVCTS TSPT | |
Greater than 97% as determined by SDS-PAGE and HPLC. |