missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TIR domain-containing adapter molecule 1 Protein

A DNA sequence encoding the Homo sapiens (Human) TIR domain-containing adapter molecule 1, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10006-iv-50ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15982738

  • 1045.00 CHF / 50 Mikrogramm


Verfügbar ab
30-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_891549.1
TIR domain-containing adapter molecule 1 Protein
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
148022
Greater than 85% as determined by SDS-PAGE.
Research Use Only
TICAM1
Recombinant Protein
Invitro-E. coli
MACTGPSLPS AFDILGAAGQ DKLLYLKHKL KTPRPGCQGQ DLLHAMVLLK LGQETEARIS LEALKADAVA RLVARQWAGV DSTEDPEEPP DVSWAVARLY HLLAEEKLCP ASLRDVAYQE AVRTLSSRDD HRLGELQDEA RNRCGWDIAG DPGSIRTLQS NLGCLPPSSA LPSGTRSLPR PIDGVSDWSQ GCSLRSTGSP ASLASNLEIS QSPTMPFLSL HRSPHGPSKL CDDPQASLVP EPVPGGCQEP EEMSWPPSGE IASPPELPSS PPPGLPEVAP DATSTGLPDT PAAPETSTNY PVECTEGSAG PQSLPLPILE PVKNPCSVKD QTPLQLSVED TTSPNTKPCP PTPTTPETSP PPPPPPPSST PCSAHLTPSS LFPSSLESSS EQKFYNFVIL HARADEHIAL RVREKLEALG VPDGATFCED FQVPGRGELS CLQDAIDHSA FIILLLTSNF DCRLSLHQVN QAMMSNLTRQ GSPDCVIPFL PLESSPAQLS SDTASLLSGL VRLDEHSQIF ARKVANTFKP HRLQARKAMW RKEQDTRALR EQSQHLDGER MQAAALNAAY SAYLQSYLSY QAQMEQLQVA FGSHMSFGTG APYGARMPFG GQVPLGAPPP FPTWPGCPQP PPLHAWQAGT PPPPSPQPAA FPQSLPFPQS PAFPTASPAP PQSPGLQPLI IHHAQMVQLG LNNHMWNQRG SQAPEDKTQE AE
Videos
Dokumentation