missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thymidylate kinase Protein
A DNA sequence encoding the Homo sapiens (Human) Thymidylate kinase, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP5955-ec-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001158503.1 | |
Thymidylate kinase Protein | |
Research Use Only | |
DTYMK | |
Recombinant Protein | |
E. coli | |
AARRGALIVL EGVDRAGKST QSRKLVEALC AAGHRAELLR FPERSTEIGK LLSSYLQKKS DVEDHSVHLL FSANRWEQVP LIKEKLSQGV TLVVDRYAFS GVAFTGAKEN FSLDWCKQPD VGLPKPDLVL FLQLQLADAA KRGAFGHERY ENGAFQERAL RCFHQLMKDT TLNWKMVDAS KSIEAVHEDI RVLSEDAIRT ATEKPLGELW K | |
Greater than 80% as determined by SDS-PAGE. |
1841 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |