missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TACI Protein

A cDNA sequence encoding the TACI was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP10873-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15965428

  • 6775.00 CHF / 1 Milligramm
Verfügbar ab: 28-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

23495
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Fully biologically active when compared to standard. The biological activity is determined by its ability to block human BAFF induced T2B cell survival using a concentration range of 1.0-3.0 microg/ml corresponding to a specific activity of 334-1000 IU/mg.
Human
Untagged
TACI protein was lyophilized from a 0.2 microm filtered concentrated solution in PBS, pH 7.4.
TACI Protein
Research Use Only
TACI
Recombinant Protein
E. coli
MSGLGRSRRG GRSRVDQEER FPQGLWTGVA MRSCPEEQYW DPLLGTCMSC KTICNHQSQR TCAAFCRSLS CRKEQGKFYD HLLRDCISCA SICGQHPKQC AYFCENKLRS PVNLPPELRR QRSGEVENNS DNSGRYQGLE HRGSEASPAL PGLKLSADQV
Greater than 95.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote