missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SYNJ2BP Protein
A cDNA sequence encoding the SYNJ2BP was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13657-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
SYNJ2BP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNGRVDY LVTEEEINLT RGPSGLGFNI VGGTDQQYVS NDSGIYVSRI KENGAAALDG RLQEGDKILS VNGQDLKNLL HQDAVDLFRN AGYAVSLRVQ HRLQVQNGPI GHRGEGDPSG | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SYNJ2BP protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |