missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Syndecan-4 / SDC4 Protein

A DNA sequence encoding the Human Syndecan-4 / SDC4, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10177-ec-100ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15982978

  • 463.00 CHF / 100 Mikrogramm


Verfügbar ab
30-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_002990.2
Syndecan-4 / SDC4 Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
ESIRETEVID PQDLLEGRYF SGALPDDEDV VGPGQESDDF ELSGSGDLDD LEDSMIGPEV VHPLVPLDNH IPERAGSGSQ VPTEPKKLEE NEVIPKRISP VEESEDVSNK VSMSSTVQGS NIFERTE
6385
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
SDC4
The specific activity is determined by binding ability in a functional ELISA. Immobilized rHuSYND4 at 500 ng/ml (100 ul/well) can bind rHubFGF with a linear range of 0.1-10 ng/ml.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Dokumentation