missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STX1A Protein
A cDNA sequence encoding the STX1A was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13632-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
6804 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKDRTQELRT AKDSDDDDDV AVTVDRDRFM DEFFEQVEEI RGFIDKIAEN VEEVKRKHSA ILASPNPDEK TKEELEELMS DIKKTANKVR SKLKSIEQSI EQEEGLNRSS ADLRIRKTQH STLSRKFVEV MSEYNATQSD YRERCKGRIQ RQLEITGRTT TSEELEDMLE SGNPAIFASG IIMDSSISKQ ALSEIETRHS EIIKLENSIR ELHDMFMDMA MLVESQ | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
STX1A Protein | |
Research Use Only | |
STX1A | |
Human | |
His | |
The protein solution contains 20mM Tris-HCl pH 7.5, 10% glycerol, and 1mM DTT. |