missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SSU72 Protein
A cDNA sequence encoding the SSU72 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13599-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
SSU72 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPSSPLR VAVVCSSNQN RSMEAHNILS KRGFSVRSFG TGTHVKLPGP APDKPNVYDF KTTYDQMYND LLRKDKELYT QNGILHMLDR NKRIKPRPER FQNCKDLFDL ILTCEERVYD QVVEDLNSRE QETCQPVHVV NVDIQDNHEE ATLGAFLICE LCQCIQHTED MENEIDELLQ EFEEKSGRTF LHTVCFY | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SSU72 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 10% glycerol. |