missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SSBP1 Protein
A cDNA sequence encoding the SSBP1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13593-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
SSBP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSESETTTS LVLERSLNRV HLLGRVGQDP VLRQVEGKNP VTIFSLATNE MWRSGDSEVY QLGDVSQKTT WHRISVFRPG LRDVAYQYVK KGSRIYLEGK IDYGEYMDKN NVRRQATTII ADNIIFLSDQ TKEKE | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SSBP1 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT. |