missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SOS1 Protein

A cDNA sequence encoding the SOS1 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13560-50ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983969

  • 1660.00 CHF / 50 Mikrogramm


Verfügbar ab
27-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

SOS1 Protein
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SOS1 protein solution (0.25 mg/ml) contains 30% glycerol, 0.1M NaCl, 1mM DTT and 0.2mM MgCl2.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
ADPEEQMRLP SADVYRFAEP DSEENIIFEE NMQPKAGIPI IKAGTVIKLI ERLTYHMYAD PNFVRTFLTT YRSFCKPQEL LSLIIERFEI PEPEPTEADR IAIENGDQPL SAELKRFRKE YIQPVQLRVL NVCRHWVEHH FYDFERDAYL LQRMEEFIGT VRGKAMKKWV ESITKIIQRK KIARDNGPGH NITFQSSPPT VEWHISRPGH IETFDLLTLH PIEIARQLTL LESDLYRAVQ PSELVGSVWT KEDKEINSPN LLKMIRHTTN LTLWFEKCIV ETENLEERVA VVSRIIEILQ VFQELNNFNG VLEVVSAMNS SPVYRLDHTF EQIPSRQKKI LEEAHELSED HYKKYLAKLR SINPPCVPFF GIYLTNILKT EEGNPEVLKR HGKELINFSK RRKVAEITGE IQQYQNQPYC LRVESDIKRF FENLNPMGNS MEKEFTDYLF NKSLEIEPRN PKPLPRFPKK YSYPLKSPGV RPSNPRPGTH HHHHH
Videos
Dokumentation