missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNTN Protein
A cDNA sequence encoding the SNTN was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13553-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
SNTN Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGGCMHSTQD KSLHLEGDPN PSAAPTSTCA PRKMPKRISI SKQLASVKAL RKCSDLEKAI ATTALIFRNS SDSDGKLEKA IAKDLLQTQF RNFAEGQETK PKYREILSEL DEHTENKLDF EDFMILLLSI TVMSDLLQNI RNVKIMK | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SNTN solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 50% glycerol and 0.15M NaCl. |