missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Small Nuclear Ribonucleoprotein, 70kDa Protein

A cDNA sequence encoding the Small Nuclear Ribonucleoprotein, 70kDa was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP14031-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15965459

  • 4920.00 CHF / 1 Milligramm
Verfügbar ab: 28-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

6625
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MTQFLPPNLL ALFAPRDPIP YLPPLEKLPH EKHHNQPYCG IAPYIREFED PRDAPPPTRA ETREERMERK RREKIERRQQ EVETELKMWD PHNDPNAQGD AFKTLFVARV NYDTTESKLR REFEVYGPIK RIHMVYSKRS GKPRGYAFIE YEHERDMHSA YKHADGKKID GRRVLVDVER GRTVKGWRPR RLGGGLGGTR RGGADVNIRH SGRDDTSRYD ERPGPSPLPH RDRDRDRERE RRERSRERDK ERERRRSRSR DRRRRSRSRD KEERRRSRER SKDKDRDRKR RSSRSRERAR RERERKEELR GGGGDMAEPS EAGDAPPDDG PPGELGPDGP DGPEEKGRDR DRERRRSHRS ERERRRDRDR DRDRDREHKR GERGSERGRD EARGGGGGQD NGLEGLGNDS RDMYMESEGG DGYLAPENGY LMEAAPE
Greater than 90% as determined by SDS-PAGE.
Small Nuclear Ribonucleoprotein, 70kDa Protein
Research Use Only
SNRNP70
Human
His
U1-snRNP is supplied in 20mM HEPES buffer pH 7.5, 0.01mM EDTA and 0.02% SDS.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote