missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SKA1 Protein
A cDNA sequence encoding the SKA1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13515-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
SKA1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASSDLE QLCSHVNEKI GNIKKTLSLR NCGQEPTLKT VLNKIGDEII VINELLNKLE LEIQYQEQTN NSLKELCESL EEDYKDIEHL KENVPSHLPQ VTVTQSCVKG SDLDPEEPIK VEEPEPVKKP PKEQRSIKEM PFITCDEFNG VPSYMKSRLT YNQINDVIKE INKAVISKYK ILHQPKKSMN SVTRNLYHRF IDEETKDTKG RYFIVEADIK EFTTLKADKK FHVLLNILRH CRRLSEVRGG GLTRYVIT | |
Greater than 80.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SKA1 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |