missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SIX1 Protein
A cDNA sequence encoding the SIX1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13513-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
SIX1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSMLPS FGFTQEQVAC VCEVLQQGGN LERLGRFLWS LPACDHLHKN ESVLKAKAVV AFHRGNFREL YKILESHQFS PHNHPKLQQL WLKAHYVEAE KLRGRPLGAV GKYRVRRKFP LPRTIWDGEE TSYCFKEKSR GVLREWYAHN PYPSPREKRE LAEATGLTTT QVSNWFKNRR QRDRAAEAKE RENTENNNSS SNKQNQLSPL EGGKPLMSSS EEEFSPPQSP DQNSVLLLQG NMGHARSSNY SLPGLTASQP SHGLQTHQHQ LQDSLLGPLT SSLVDLGS | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SIX1 solution contains 20mM Tris-HCl buffer (pH 8.0), 2M Urea and 10% glycerol. |