missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SH3BGRL2 Protein

A cDNA sequence encoding the SH3BGRL2 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13488-20ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983729

  • 188.00 CHF / 20 Mikrogramm


Verfügbar ab
27-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

SH3BGRL2 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The SH3BGRL2 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMVIRVFI ASSSGFVAIK KKQQDVVRFL EANKIEFEEV DITMSEEQRQ WMYKNVPPEK KPTQGNPLPP QIFNGDRYCG DYDSFFESKE SNTVFSFLGL KPRLASKAEP
Videos
Dokumentation