missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SerpinI1 / Neuroserpin Protein

A DNA sequence encoding the Human SerpinI1 / Neuroserpin, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10160-ec-25ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15982938

  • 313.00 CHF / 25 Mikrogramm


Verfügbar ab
28-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_001116224.1
SerpinI1 / Neuroserpin Protein
25 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
TGATFPEEAI ADLSVNMYNR LRATGEDENI LFSPLSIALA MGMMELGAQG STQKEIRHS MGYDSLKNGE EFSFLKEFSN MVTAKESQYV MKIANSLFVQ NGFHVNEEF LQMMKKYFNA AVNHVDFSQN VAVANYINKW VENNTNNLVK DLVSPRDFD AATYLALINA VYFKGNWKSQ FRPENTRTFS FTKDDESEVQ IPMMYQQGE FYYGEFSDGS NEAGGIYQVL EIPYEGDEIS MMLVLSRQEV PLATLEPLV KAQLVEEWAN SVKKQKVEVY LPRFTVEQEI DLKDVLKALG ITEIFIKDA NLTGLSDNKE IFLSKAIHKS FLEVNEEGSE AAAVSGMIAI SRMAVLYPQ VIVDHPFFFL IRNRRTGTIL FMGRVMHPET MNTSGHD
5274
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
SERPINI1
The ED50 as determined by a cell proliferation assay using rat C6 cells is less than 0.5 ug/ml, corresponding to a specific activity of > 2000 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.5, lyophilized
Videos
Dokumentation