missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SERPINA1 Protein

A cDNA sequence encoding the SERPINA1 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP10864-100ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15965398

Menge Preis
1 192.32 CHF / 100 Mikrogramm
6 177.22 CHF / 100 Mikrogramm
Voraussichtliches Versanddatum: 29-10-2021

Zum Warenkorb hinzufügen


Beschreibung

Beschreibung

Spezifikation

Spezifikation

SERPINA1 Protein
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Rice
MPSSVSWGIL LLAGLCCLVP VSLAEDPQGD AAQKTDTSHH DQDHPTFNKI TPNLAEFAFS LYRQLAHQSN STNIFFSPVS IATAFAMLSL GTKADTHDEI LEGLNFNLTE IPEAQIHEGF QELLRTLNQP DSQLQLTTGN GLFLSEGLKL VDKFLEDVKK LYHSEAFTVN FGDTEEAKKQ INDYVEKGTQ GKIVDLVKEL DRDTVFALVN YIFFKGKWER PFEVKDTEEE DFHVDQVTTV KVPMMKRLGM FNIQHCKKLS SWVLLMKYLG NATAIFFLPD EGKLQHLENE LTHDIITKFL ENEDRRSASL HLPKLSITGT YDLKSVLGQL GITKVFSNGA DLSGVTEEAP LKLSKAVHKA VLTIDEKGTE AAGAMFLEAI PMSIPPEVKF NKPFVFLMIE QNTKSPLFMG KVVNPTQK
Greater than 95% as determined by SDS-PAGE.
Research Use Only
3∼5 mg SERPINA1 will inhibit 1 mg PPE with an activity of 10.8 units per mg protein.
Human
Untagged
SERPINA1 was lyophilized from a concentrated solution containing recombinant Albumin.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate