missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SEPX1 Protein
A cDNA sequence encoding the SEPX1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13453-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
SEPX1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSFCSFFGGE VFQNHFEPGV YVCAKCGYEL FSSRSKYAHS SPWPAFTETI HADSVAKRPE HNRSEALKVS CGKCGNGLGH EFLNDGPKPG QSRFCIFSSS LKFVPKGKET SASQGH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SEPX1 solution (0.5 mg/ml) contains 20mM Tris-HCl Buffer (pH 7.5), 1mM DTT, 0.1mM PMSF, 2mM EDTA and 10% Glycerol. |