missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SDR16C5 Protein

A cDNA sequence encoding the SDR16C5 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13443-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15963579

  • 3817.53 CHF / 1 Milligramm


Voraussichtliches Versanddatum: 10-11-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

SDR16C5 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SDR16C5 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSPKPRKNV AGEIVLITGA GSGLGRLLAL QFARLGSVLV LWDINKEGNE ETCKMAREAG ATRVHAYTCD CSQKEGVYRV ADQVKKEVGD VSILINNAGI VTGKKFLDCP DELMEKSFDV NFKAHLWTYK AFLPAMIAND HGHLVCISSS AGLSGVNGLA DYCASKFAAF GFAESVFVET FVQKQKGIKT TIVCPFFIKT GMFEGCTTGC PSLLPILEPK YAVEKIVEAI LQEKMYLYMP K
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate