missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SCGB1D1 Protein

A cDNA sequence encoding the SCGB1D1 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13428-10ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983529

  • 188.00 CHF / 10 Mikrogramm


Verfügbar ab
29-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

SCGB1D1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SCGB1D1 filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 20mM Tris buffer and 50mM NaCl, pH 7.5.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHAS VVCQALGSEI TGFLLAGKPV FKFQLAKFKA PLEAVAAKME VKKCVDTMAY EKRVLITKTL GKIAEKCDR
Videos
Dokumentation