missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A8 Protein
A cDNA sequence encoding the S100A8 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 5210.00 CHF
Spezifikation
Name | S100A8 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15993439
|
enQuireBio™
QP13399-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 14-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15983439
|
enQuireBio™
QP13399-1MG |
1 mg |
5210.00 CHF
1 Milligramm |
Verfügbar ab: 14-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15903449
|
enQuireBio™
QP13399-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 14-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
S100A8 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
S100A8 was filtered (0.4microm) and lyophilized in 0.5 mg/ml in 20mM Tris and 100mM NaCl, pH 7.5. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS MLTELEKALN SIIDVYHKYS LIKGNFHAVY RDDLKKLLET ECPQYIRKKG ADVWFKELDI NTDGAVNFQEMLTELEKALN SIIDVYHKYS LIKGNFHAVY RDDLKKLLET ECPQYIRKKG ADVWFKELDI NTDGAVNFQE FLILVIKMGV AAHKKSHEES HKE |