missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A13 Protein
A cDNA sequence encoding the S100A13 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13389-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
S100A13 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAAEPLTELE ESIETVVTTF FTFARQEGRK DSLSVNEFKE LVTQQLPHLL KDVGSLDEKM KSLDVNQDSE LKFNEYWRLI GELAKEIRKK KDLKIRKK | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
S100A13 protein at 1 mg/ml in 20mM Tris-HCL, pH 8.0, 50mM NaCl and 20% Glycerol. |