missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RWDD1 Protein

A cDNA sequence encoding the RWDD1 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13376-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983369

  • 7525.00 CHF / 1 Milligramm
Verfügbar ab: 28-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

RWDD1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMTDYGEE QRNELEALES IYPDSFTVLS ENPPSFTITV TSEAGENDET VQTTLKFTYS EKYPDEAPLY EIFSQENLED NDVSDILKLL ALQAEENLGM VMIFTLVTAV QEKLNEIVDQ IKTRREEEKK QKEKEAEEAE KQLFHGTPVT IENFLNWKAK FDAELLEIKK KRMKEEEQAG KNKLSGKQLF ETDHNLDTSD IQFLEDAGNN VEVDESLFQE MDDLELEDDE DDPDYNPADP ESDSAD
Greater than 85.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RWDD1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 2mM DTT.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote