missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RSPO3 Protein

A cDNA sequence encoding the RSPO3 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13366-20ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983339

  • 188.00 CHF / 20 Mikrogramm


Verfügbar ab
28-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

RSPO3 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RSPO3 was lyophilized from a 0.2microm filtered solution in PBS, pH 7.4.
Greater than 95% as determined by SDS PAGE.
Research Use Only
Recombinant Protein
HEK 293
QNASRGRRQR RMHPNVSQGC QGGCATCSDY NGCLSCKPRL FFALERIGMK QIGVCLSSCP SGYYGTRYPD INKCTKCKAD CDTCFNKNFC TKCKSGFYLH LGKCLDNCPE GLEANNHTME CVSIVHCEVS EWNPWSPCTK KGKTCGFKRG TETRVREIIQ HPSAKGNLCP PTNETRKCTV DDIEGRMDEP KSCDKTHTCP PCPAPELLGG PSVFLFPPKP KDTLMISRTP EVTCVVVDVS HEDPEVKFNW YVDGVEVHNA KTKPREEQYN STYRVVSVLT VLHQDWLNGK EYKCKVSNKA LPAPIEKTIS KAKGQPREPQ VYTLPPSREE MTKNQVSLTC LVKGFYPSDI AVEWESNGQP ENNYKTTPPV LDSDGSFFLY SKLTVDKSRW QQGNVFSCSV MHEALHNHYT QKSLSLSPGK HHHHHH
Videos
Dokumentation