missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RPS2 Protein

A cDNA sequence encoding the RPS2 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13354-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983299

  • 3735.00 CHF / 1 Milligramm


Verfügbar ab
28-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

RPS2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RPS2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0),10% glycerol and 0.4M Urea.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMADDAGA AGGPGGPGGP GMGNRGGFRG GFGSGIRGRG RGRGRGRGRG RGARGGKAED KEWMPVTKLG RLVKDMKIKS LEEIYLFSLP IKESEIIDFF LGASLKDEVL KIMPVQKQTR AGQRTRFKAF VAIGDYNGHV GLGVKCSKEV ATAIRGAIIL AKLSIVPVRR GYWGNKIGKP HTVPCKVTGR CGSVLVRLIP APRGTGIVSA PVPKKLLMMA GIDDCYTSAR GCTATLGNFA KATFDAISKT YSYLTPDLWK ETVFTKSPYQ EFTDHLVKTH TRVSVQRTQA PAVATT
Videos
Dokumentation