missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPS19 Protein
A cDNA sequence encoding the RPS19 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13353-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
6223 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPGVTVK DVNQQEFVRA LAAFLKKSGK LKVPEWVDTV KLAKHKELAP YDENWFYTRA ASTARHLYLR GGAGVGSMTK IYGGRQRNGV MPSHFSRGSK SVARRVLQAL EGLKMVEKDQ DGGRKLTPQG QRDLDRIAGQ VAAANKKH | |
Greater than 85% as determined by SDS-PAGE. |
RPS19 Protein | |
Research Use Only | |
RPS19 | |
Human | |
His | |
The RPS19 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) containing 0.2M NaCl, 40% glycerol and 2mM DTT. |