missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RPL34 Protein

A cDNA sequence encoding the RPL34 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13347-20ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983279

  • 188.00 CHF / 20 Mikrogramm


Verfügbar ab
26-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

RPL34 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The RPL34 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMVQRLTY RRRLSYNTAS NKTRLSRTPG NRIVYLYTKK VGKAPKSACG VCPGRLRGVR AVRPKVLMRL SKTKKHVSRA YGGSMCAKCV RDRIKRAFLI EEQKIVVKVL KAQAQSQKAK
Videos
Dokumentation