missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPIA Protein
A cDNA sequence encoding the RPIA was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Gen-ID (Entrez) | 22934 |
---|---|
Name | RPIA Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gensymbol | RPIA |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15983259
|
enQuireBio™
QP13340-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15963259
|
enQuireBio™
QP13340-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15973259
|
enQuireBio™
QP13340-1MG |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
22934 | |
Research Use Only | |
RPIA | |
Human | |
His | |
The RPIA solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT,40% glycerol, 200mM NaCl, 2mM EDTA and 0.2mM PMSF. |
RPIA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MQRPGPFSTL YGRVLAPLPG RAGGAASGGG GNSWDLPGSH VRLPGRAQSG TRGGAGNTST SCGDSNSICP APSTMSKAEE AKKLAGRAAV ENHVRNNQVL GIGSGSTIVH AVQRIAERVK QENLNLVCIP TSFQARQLIL QYGLTLSDLD RHPEIDLAID GADEVDADLN LIKGGGGCLT QEKIVAGYAS RFIVIADFRK DSKNLGDQWH KGIPIEVIPM AYVPVSRAVS QKFGGVVELR MAVNKAGPVV TDNGNFILDW KFDRVHKWSE VNTAIKMIPG VVDTGLFINM AERVYFGMQD GSVNMREKPF C | |
Greater than 90% as determined by SDS-PAGE. |