missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RBP5 Protein

A cDNA sequence encoding the RBP5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13284-2ug

Additional Details : Gewicht : 0.01000kg

 View more versions of this product

Product Code. 15983079

  • 125.00 CHF / 2 Mikrogramm


Verfügbar ab
28-10-2020
Add to basket

Specifications

Specifications

RBP5 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The RBP5 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 2mM DTT and 100mM NaCl.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MPPNLTGYYR FVSQKNMEDY LQALNISLAV RKIALLLKPD KEIEHQGNHM TVRTLSTFRN YTVQFDVGVE FEEDLRSVDG RKCQTIVTWE EEHLVCVQKG EVPNRGWRHW LEGEMLYLEL TARDAVCEQV FRKVR
Videos
Documents