missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RBM11 Protein
A cDNA sequence encoding the RBM11 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Name | RBM11 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15983059
|
enQuireBio™
QP13278-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15973059
|
enQuireBio™
QP13278-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15963059
|
enQuireBio™
QP13278-1MG |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
RBM11 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RBM11 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMFPAQEE ADRTVFVGNL EARVREEILY ELFLQAGPLT KVTICKDREG KPKSFGFVCF KHPESVSYAI ALLNGIRLYG RPINVQYRFG SSRSSEPANQ SFESCVKINS HNYRNEEMLV GRSSFPMQYF PINNTSLPQE YFLFQKMQWH VYNPVLQLPY YEMTAPLPNS ASVSSSLNHV PDLEAGPSSY KWTHQQPSDS DLYQMTAPLP NSASVSSSLN HVPDLEAGPS SYKWTHQQPS DSDLYQMNKR KRQKQTSDSD SSTDNNRGNE CSQKFRKSKK KKRY | |
Greater than 85% as determined by SDS-PAGE. |