missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RASD2 Protein

A cDNA sequence encoding the RASD2 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13271-20ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15963039

Menge Preis
1 192.32 CHF / 20 Mikrogramm
6 177.22 CHF / 20 Mikrogramm
Voraussichtliches Versanddatum: 28-10-2021

Zum Warenkorb hinzufügen


Beschreibung

Beschreibung

Spezifikation

Spezifikation

RASD2 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RASD2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMMKTLSS GNCTLSVPAK NSYRMVVLGA SRVGKSSIVS RFLNGRFEDQ YTPTIEDFHR KVYNIRGDMY QLDILDTSGN HPFPAMRRLS ILTGDVFILV FSLDNRESFD EVKRLQKQIL EVKSCLKNKT KEAAELPMVI CGNKNDHGEL CRQVPTTEAE LLVSGDENCA YFEVSAKKNT NVDEMFYVLF SMAKLPHEMS PALHRKISVQ YGDAFHPRPF CMRRVKEMDA YGMVSPFARR PSVNSDLKYI KAKVLREGQA RERDKCTIQ
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate