missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAP1A Protein
A cDNA sequence encoding the RAP1A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13265-10ug
Detalles adicionales : Gewicht : 0.01000kg
Especificaciones
RAP1A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MREYKLVVLG SGGVGKSALT VQFVQGIFVE KYDPTIEDSY RKQVEVDCQQ CMLEILDTAG TEQFTAMRDL YMKNGQGFAL VYSITAQSTF NDLQDLREQI LRVKDTEDVP MILVGNKCDL EDERVVGKEQ GQNLARQWCN CAFLESSAKS KINVNEIFYD LVRQINRKTP VEKKKPKKKS C | |
Greater than 85.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
0.25 mg/ml solution containing 20mM Tris HCL pH 8.0, 5mM DTT, 0.2M NaCl and 40% glycerol. |