missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RANBP1 Protein

A cDNA sequence encoding the RANBP1 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13261-25ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15963009

  • 203.00 CHF / 25 Mikrogramm
Verfügbar ab: 21-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

RANBP1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMAAAKD THEDHDTSTE NTDESNHDPQ FEPIVSLPEQ EIKTLEEDEE ELFKMRAKLF RFASENDLPE WKERGTGDVK LLKHKEKGAI RLLMRRDKTL KICANHYITP MMELKPNAGS DRAWVWNTHA DFADECPKPE LLAIRFLNAE NAQKFKTKFE ECRKEIEERE KKAGSGKNDH AEKVAEKLEA LSVKEETKED AEEKQ
Greater than 95% as determined by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The RANBP1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 10% glycerol.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote