missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB6B Protein
A cDNA sequence encoding the RAB6B was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13245-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
RAB6B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSAGGDFGNP LRKFKLVFLG EQSVGKTSLI TRFMYDSFDN TYQATIGIDF LSKTMYLEDR TVRLQLWDTA GQERFRSLIP SYIRDSTVAV VVYDITNLNS FQQTSKWIDD VRTERGSDVI IMLVGNKTDL ADKRQITIEE GEQRAKELSV MFIETSAKTG YNVKQLFRRV ASALPGMENV QEKSKEGMID IKLDKPQEPP ASEGGCSC | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RAB6B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |