missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB35 Protein
A cDNA sequence encoding the RAB35 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13239-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
RAB35 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MARDYDHLFK LLIIGDSGVG KSSLLLRFAD NTFSGSYITT IGVDFKIRTV EINGEKVKLQ IWDTAGQERF RTITSTYYRG THGVIVVYDV TSAESFVNVK RWLHEINQNC DDVCRILVGN KNDDPERKVV ETEDAYKFAG QMGIQLFETS AKENVNVEEM FNCITELVLR AKKDNLAKQQ QQQQNDVVKL TKNSKRKKRC C | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RAB35 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 40% glycerol, 0.15M NaCl and 1mM DTT. |