missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human QTRT1 Protein
A cDNA sequence encoding the QTRT1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13226-50ug
Additional Details : Gewicht : 0.01000kg
Specifications
QTRT1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEFMAGAA TQASLESAPR IMRLVAECSR SRARAGELWL PHGTVATPVF MPVGTQATMK GITTEQLDAL GCRICLGNTY HLGLRPGPEL IQKANGLHGF MNWPHNLLTD SGGFQMVSLV SLSEVTEEGV RFRSPYDGNE TLLSPEKSVQ IQNALGSDII MQLDDVVSST VTGPRVEEAM YRSIRWLDRC IAAHQRPDKQ NLFAIIQGGL DADLRATCLE EMTKRDVPGF AIGGLSGGES KSQFWRMVAL STSRLPKDKP RYLMGVGYAT DLVVCVALGC DMFDCVFPTR TARFGSALVP TGNLQLRKKV FEKDFGPIDP ECTCPTCQKH SRAFLHALLH SDNTAALHHL TVHNIAYQLQ LMSAVRTSIV EKRFPDFVRD FMGAMYGDPT LCPTWATDAL ASVGITLG | |
Greater than 80.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
QTRT1 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 30% glycerol and 1mM DTT. |