missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human QTRT1 Protein
A cDNA sequence encoding the QTRT1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 1735.00 CHF
Spezifikation
Name | QTRT1 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15972899
|
enQuireBio™
QP13226-1UG |
1 μg |
131.00 CHF
1 Mikrogramm |
Verfügbar ab: 27-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15992899
|
enQuireBio™
QP13226-5UG |
5 μg |
203.00 CHF
5 Mikrogramm |
Verfügbar ab: 27-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15982899
|
enQuireBio™
QP13226-50UG |
50 μg |
1735.00 CHF
50 Mikrogramm |
Verfügbar ab: 27-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
QTRT1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
QTRT1 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 30% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEFMAGAA TQASLESAPR IMRLVAECSR SRARAGELWL PHGTVATPVF MPVGTQATMK GITTEQLDAL GCRICLGNTY HLGLRPGPEL IQKANGLHGF MNWPHNLLTD SGGFQMVSLV SLSEVTEEGV RFRSPYDGNE TLLSPEKSVQ IQNALGSDII MQLDDVVSST VTGPRVEEAM YRSIRWLDRC IAAHQRPDKQ NLFAIIQGGL DADLRATCLE EMTKRDVPGF AIGGLSGGES KSQFWRMVAL STSRLPKDKP RYLMGVGYAT DLVVCVALGC DMFDCVFPTR TARFGSALVP TGNLQLRKKV FEKDFGPIDP ECTCPTCQKH SRAFLHALLH SDNTAALHHL TVHNIAYQLQ LMSAVRTSIV EKRFPDFVRD FMGAMYGDPT LCPTWATDAL ASVGITLG | |
Greater than 80.0% as determined by SDS-PAGE. |