missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTP4A3 Protein

A cDNA sequence encoding the PTP4A3 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13210-10ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15962849

Menge Preis
1 128.22 CHF / 10 Mikrogramm
10 117.77 CHF / 10 Mikrogramm
Voraussichtliches Versanddatum: 03-11-2021

Zum Warenkorb hinzufügen


Beschreibung

Beschreibung

Spezifikation

Spezifikation

PTP4A3 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PTP4A3 solution contains 20mM Tris-HCl buffer (pH 8.0), 2mM EDTA, 1mM DTT and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MARMNRPAPV EVSYKHMRFL ITHNPTNATL STFIEDLKKY GATTVVRVCE VTYDKTPLEK DGITVVDWPF DDGAPPPGKV VEDWLSLVKA KFCEAPGSCV AVHCVAGLGR APVLVALALI ESGMKYEDAI QFIRQKRRGA INSKQLTYLE KYRPKQRLRFKDPHTHKTRC CVM
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate