missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTP4A2 Protein

A cDNA sequence encoding the PTP4A2 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13209-20ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15982839

  • 188.00 CHF / 20 Mikrogramm


Verfügbar ab
28-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

PTP4A2 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PTP4A2 (0.5 mg/ml) protein solution contains 20mM Tris-HCl, pH 8.0, 1mM EDTA and 10% Glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMNRP APVEISYENM RFLITHNPTN ATLNKFTEEL KKYGVTTLVR VCDATYDKAPVEKEGIHVLD WPFDDGAPPP NQIVDDWLNL LKTKFREEPG CCVAVHCVAG LGRAPVLVAL ALIECGMKYE DAVQFIRQKR RGAFNSKQLL YLEKYRPKMR LRFRDTNGHC CVQ
Videos
Dokumentation