missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTP4A2 Protein
A cDNA sequence encoding the PTP4A2 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Name | PTP4A2 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15992839
|
enQuireBio™
QP13209-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15982839
|
enQuireBio™
QP13209-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15972839
|
enQuireBio™
QP13209-1MG |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PTP4A2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PTP4A2 (0.5 mg/ml) protein solution contains 20mM Tris-HCl, pH 8.0, 1mM EDTA and 10% Glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMNRP APVEISYENM RFLITHNPTN ATLNKFTEEL KKYGVTTLVR VCDATYDKAPVEKEGIHVLD WPFDDGAPPP NQIVDDWLNL LKTKFREEPG CCVAVHCVAG LGRAPVLVAL ALIECGMKYE DAVQFIRQKR RGAFNSKQLL YLEKYRPKMR LRFRDTNGHC CVQ | |
Greater than 90.0% as determined by SDS-PAGE. |