missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein CYR61 Protein

A DNA sequence encoding the Human Protein CYR61, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10401-ec-250ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983738

  • 2340.00 CHF / 250 Mikrogramm


Verfügbar ab
28-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_001545.2
Protein CYR61 Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
TCPAACHCPL EAPKCAPGVG LVRDGCGCCK VCAKQLNEDC SKTQPCDHTK GLECNFGASS TALKGICRAQ SEGRPCEYNS RIYQNGESFQ PNCKHQCTCI DGAVGCIPLC PQELSLPNLG CPNPRLVKVT GQCCEEWVCD EDSIKDPMED QDGLLGKELG FDASEVELTR NNELIAVGKG SSLKRLPVFG MEPRILYNPL QGQKCIVQTT SWSQCSKTCG TGISTRVTND NPECRLVKET RICEVRPCGQ PVYSSLKKGK KCSKTKKSPE PVRFTYAGCL SVKKYRPKYC GSCVDGRCCT PQLTRTVKMR FRCEDGETFS KNVMMIQSCK CNYNCPHANE AAFPFYRLFN DIHKFRD
3491
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
CYR61
The ED50 as determined by a cell proliferation assay using murine Balb/3T3 cells is less than 3.0 ug/ml, corresponding to a specific activity of > 330 IU/mg.
Human
Untagged
0.2 um filtered citrate buffer solution, 300 mM NaCl, pH 3.0, lyophilized
Videos
Dokumentation