missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Procalcitonin Protein
A cDNA sequence encoding the Procalcitonin was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13155-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
Procalcitonin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
PCT is supplied in 20mM Tris-HCl pH 8.0 and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Greater than 95.0% as determined by SDS-PAGE. |