missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Procalcitonin Protein
A cDNA sequence encoding the Procalcitonin was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10847-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
Procalcitonin Protein | |
Research Use Only | |
Human Procalcitonin was tested using Biomerieux reagents on the Min-Vidas. The assay was run at 80ng/ml with a typical recovery of 80 percent. | |
Human | |
Untagged | |
Lyophilized from 10mM sodium phosphate pH 7.5. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
APFRSALESS PADPATLSED EARLLLAALV QDYVQMKASE LEQEQEREGS SLDSPRSKRC GNLSTCMLGT YTQDFNKFHT FPQTAIGVGA PGKKRDMSSD LERDHRPHVS MPQNAN | |
Greater than 95.0% as determined by SDS-PAGE. |